Merck
  • Improved pruritus correlates with lower levels of IL-31 in CTCL patients under different therapeutic modalities.
Improved pruritus correlates with lower levels of IL-31 in CTCL patients under different therapeutic modalities.
Clinical immunology (Orlando, Fla.) (2015-03-13)
Filiberto Cedeno-Laurent, Elisha M Singer, Maria Wysocka, Bernice M Benoit, Carmela C Vittorio, Ellen J Kim, Gil Yosipovitch, Alain H Rook
ZUSAMMENFASSUNG
Pruritus is one of the cardinal symptoms found in patients with leukemic cutaneous T cell lymphoma (CTCL). The nature of the pruritus experienced by CTCL patients is complex, involving different pathways and cell mediators, thus making it poorly responsive to conventional anti-itch therapies. Recent reports highlight the role of interleukin 31 (IL-31) as a novel cytokine involved in the pathogenesis of pruritus in atopic dermatitis and CTCL. Here we provide both in vivo and in vitro evidence suggesting that histone deacetylase (HDAC) inhibitors may mitigate itch through lowering of levels of IL-31-expressing T cells. Furthermore, we demonstrate that chemokine receptor type-4 (CCR4)-bearing T cells are a main source of IL-31 in CTCL, and that neutralizing the IL-31 pathway through targeting of the CCR4-expressing T cells may represent a promising therapeutic strategy for symptomatic relief in CTCL.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, ≥99% (HPLC and TLC), BioXtra, Molecular Biology
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Brefeldin A aus Penicillium brefeldianum, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO