Merck
Chitosan/mangiferin particles for Cr(VI) reduction and removal.
International journal of biological macromolecules (2015-04-07)
Caroline de G Sampaio, Lucas S Frota, Herbert S Magalhães, Lilian M U Dutra, Danilo C Queiroz, Rinaldo S Araújo, Helena Becker, José R R de Souza, Nágila M P S Ricardo, Maria T S Trevisan
ZUSAMMENFASSUNG
In this work, chitosan/mangiferin particles (CMP) were prepared by spray-drying technique and characterized by SEM, DLS, FTIR, HPLC-UV and adsorption studies to investigate a possible application as a preventive material in cases of human and animal contamination with Cr(VI). CMP presented sizes ranging from nano to micrometers. Chitosan and mangiferin (MA) presence in the powder was confirmed by FTIR and MA quantification (136 μg/mg) was performed using a calibration curve prepared by HPLC-UV. Adsorption capacity of Cr(VI) onto CMP was compared with chitosan and investigated in a batch system by considering the effects of various parameters like contact time, initial concentration of adsorbent and pH. Cr(VI) removal is pH dependent and it was found to be maximum at pH 5.0. The results showed that CMP has a potential application as a preventive material in cases of human or animal contamination with Cr(VI).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,5-Diphenylcarbazid, ACS reagent
Sigma-Aldrich
Cytidin 5′-Monophosphat, Sigma Grade, ≥99% (HPLC), synthetic, powder
Sigma-Aldrich
1,5-Diphenylcarbazid, reagent grade