Merck
  • Stabilization of dengue virus polymerase in de novo initiation assay provides advantages for compound screening.
Stabilization of dengue virus polymerase in de novo initiation assay provides advantages for compound screening.
Antiviral research (2015-04-22)
Pornwaratt Niyomrattanakit, Kah Fei Wan, Ka Yan Chung, Siti Nurdiana Abas, Cheah Chen Seh, Hongping Dong, Chin Chin Lim, Alexander Theodore Chao, Chang Bok Lee, Shahul Nilar, Julien Lescar, Pei-Yong Shi, David Beer, Siew Pheng Lim
ZUSAMMENFASSUNG
Dengue virus (DENV) NS5 protein comprises an N-terminal methyltransferase domain and a C-terminal RNA-dependent RNA polymerase domain (RdRp). DENV RdRp is responsible for viral RNA synthesis via a de novo initiation mechanism and represents an attractive target for anti-viral therapy. Herein we describe the characterization of its de novo initiation activities by PAGE analyses and the knowledge gained was used to develop a fluorescent-based assay. A highly processive and robust assay was achieved by addition of cysteine in the assay buffer. This stabilized the apo-enzyme, and rendered optimal de novo initiation activity while balancing its intrinsic terminal transferase activity. Steady-state kinetic parameters of the NTP and RNA substrates under these optimal conditions were determined for DENV1-4 FL NS5. Heavy metal ions such as Zn(++) and Co(++) as well as high levels of monovalent salts, suppressed DENV polymerase de novo initiation activities. This assay was validated with nucleotide chain terminators and used to screen two diverse small library sets. The screen data obtained was further compared with concurrent screens performed with a DENV polymerase elongation fluorescent assay utilizing pre-complexed enzyme-RNA. A higher hit-rate was obtained for the de novo initiation assay compared to the elongation assay (∼2% versus ∼0.1%). All the hits from the latter assay are also identified in the de novo initiation assay, indicating that the de novo initiation assay performed with the stabilized apo-enzyme has the advantage of providing additional chemical starting entities for inhibiting this enzyme.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
N-Methyldiethanolamin, ≥99%
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
DL-Cystein, technical grade
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C