Merck
  • Fabrication of fibrous amidoxime-functionalized mesoporous silica microsphere and its selectively adsorption property for Pb(2+) in aqueous solution.
Fabrication of fibrous amidoxime-functionalized mesoporous silica microsphere and its selectively adsorption property for Pb(2+) in aqueous solution.
Journal of hazardous materials (2015-05-06)
Yunyun Xie, Jie Wang, Mozhen Wang, Xuewu Ge
ZUSAMMENFASSUNG
Fibrous cyano-modified mesoporous SiO2 microspheres with specific surface area of ca. 300 m(2) g(-1) have been successfully fabricated respectively by in-situ synthesis and post-modification methods, based on the hydrolysis of ethyl silicate in the presence of (2-cyanoethyl)triethoxysilane at a certain condition. TEM observations show that the average diameters of the prepared fibrous cyano-modified SiO2 microspheres by these two methods are 68 and 211 nm, respectively. The N2 adsorption-desorption isotherms analysis on the fibrous SiO2 microspheres show sharp peaks in the 10-20 nm range. After the cyano groups transformed to amidoxime groups, the adsorption behavior of the fibrous amidoxime-functionalized mesoporous SiO2 microspheres for Fe(3+), Cu(2+), and Pb(2+) was investigated. The results show that the prepared SiO2 microspheres can selectively adsorb Pb(2+). The maximum equilibrium adsorption capacity for Pb(2+) could reach 284 mg/g. The desorption of Pb(2+) in 2M HNO3 completes within 60 min. The efficiency of the desorption is as high as 96.2%. This work provides the methods to prepare amidoxime-functionalized SiO2 microsphere with high specific surface area and total pore volume, which has the potential to be applied as an efficient adsorbent for specific heavy metal ions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Natriumcarbonat, powder, 99.999% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
Sigma-Aldrich
Cyclohexan, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Natriumcarbonat, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O