Merck
Influence of deep-frying using various commercial oils on acrylamide formation in French fries.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-05-09)
Hao Zhang, Hui Zhang, Lilin Cheng, Li Wang, Haifeng Qian
ZUSAMMENFASSUNG
This study investigated the effect of different types of commercial oils (rice bran oil, shortening oil, high-oleic rapeseed oil, low-erucic acid rapeseed oil, blend oil A and blend oil B) and frying cycles on acrylamide formation during the preparation of French fries by deep-frying. Frying was carried out in intermittent mode (two batches each for 12 min without any time lag) and repeated for 600 frying cycles. Results indicated that the French fries that were fried in oils having lower heat transfer coefficients contained lower acrylamide concentrations (913 µg kg(-1)), whereas those fried with oils having higher heat transfer coefficients contained higher acrylamide concentrations (1219 µg kg(-1)). Unlike the peroxide value, acrylamide levels in French fries did not change significantly with an increase in the number of frying cycles when tested for 600 frying cycles for every type of oil. This study clearly indicates that the contribution of frying oils to the formation of acrylamide should not be neglected due to their different heat transfer coefficients. On the other hand, continuous use of frying oil does not lead to a higher acrylamide concentration in French fries.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.