Merck
  • Improved treatment of systemic blood infections using antibiotics with extracorporeal opsonin hemoadsorption.
Improved treatment of systemic blood infections using antibiotics with extracorporeal opsonin hemoadsorption.
Biomaterials (2015-08-09)
Tohid F Didar, Mark J Cartwright, Martin Rottman, Amanda R Graveline, Nazita Gamini, Alexander L Watters, Daniel C Leslie, Tadanori Mammoto, Melissa J Rodas, Joo H Kang, Anna Waterhouse, Benjamin T Seiler, Patrick Lombardo, Elisabet I Qendro, Michael Super, Donald E Ingber
ZUSAMMENFASSUNG
Here we describe development of an extracorporeal hemoadsorption device for sepsis therapy that employs commercially available polysulfone or polyethersulfone hollow fiber filters similar to those used clinically for hemodialysis, covalently coated with a genetically engineered form of the human opsonin Mannose Binding Lectin linked to an Fc domain (FcMBL) that can cleanse a broad range of pathogens and endotoxin from flowing blood without having to first determine their identity. When tested with human whole blood in vitro, the FcMBL hemoadsorption filter (FcMBL-HF) produced efficient (90-99%) removal of Gram negative (Escherichia coli) and positive (Staphylococcus aureus) bacteria, fungi (Candida albicans) and lipopolysaccharide (LPS)-endotoxin. When tested in rats, extracorporeal therapy with the FcMBL-HF device reduced circulating pathogen and endotoxin levels by more than 99%, and prevented pathogen engraftment and inflammatory cell recruitment in the spleen, lung, liver and kidney when compared to controls. Studies in rats revealed that treatment with bacteriocidal antibiotics resulted in a major increase in the release of microbial fragments or 'pathogen-associated molecular patterns' (PAMPs) in vivo, and that these PAMPs were efficiently removed from blood within 2 h using the FcMBL-HF; in contrast, they remained at high levels in animals treated with antibiotics alone. Importantly, cleansing of PAMPs from the blood of antibiotic-treated animals with the FcMBL-hemoadsorbent device resulted in reduced organ pathogen and endotoxin loads, suppressed inflammatory responses, and resulted in more stable vital signs compared to treatment with antibiotics alone. As PAMPs trigger the cytokine cascades that lead to development of systemic inflammatory response syndrome and contribute to septic shock and death, co-administration of FcMBL-hemoadsorption with antibiotics could offer a more effective approach to sepsis therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98.0% (NT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, tablet, 1 mg substrate per tablet