Merck
  • Simple synthesis of Prussian blue analogues in room temperature ionic liquid solution and their catalytic application in epoxidation of styrene.
Simple synthesis of Prussian blue analogues in room temperature ionic liquid solution and their catalytic application in epoxidation of styrene.
Dalton transactions (Cambridge, England : 2003) (2015-06-24)
Qianqian Wang, Ning Wang, Sifa He, Jihua Zhao, Jian Fang, Weiguo Shen
ZUSAMMENFASSUNG
In this article, a new application of the room temperature ionic liquid, tetramethylammonium tetrafluoroborate, is described. This ionic liquid is used as a stabilizing agent and the reaction medium for the synthesis of CoFe Prussian blue analogue nanoparticles using N,N-dimethyl formamide as a complexing agent. The as-synthesized Prussian blue analogues were characterized by various techniques and were used to catalyze the reaction of epoxidation of styrene. The catalytic activity of Prussian blue analogues prepared in ionic liquid was superior to that of Prussian blue analogues prepared in aqueous solution.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formamid, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Formamid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Cobalt(II)-nitrat Hexahydrat, 99.999% trace metals basis
Sigma-Aldrich
Formamid -Lösung, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamid -Lösung, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.