Merck
  • Material Properties and Antimicrobial Activity of Polyhydroxybutyrate (PHB) Films Incorporated with Vanillin.
Material Properties and Antimicrobial Activity of Polyhydroxybutyrate (PHB) Films Incorporated with Vanillin.
Applied biochemistry and biotechnology (2015-05-24)
Janifer Raj Xavier, Sudalaimuthu Thangaraj Babusha, Johnsy George, Karna Venkata Ramana
ZUSAMMENFASSUNG
Polyhydroxybutyrate (PHB) was produced by Bacillus mycoides DFC 1, isolated from garden soil. Antimicrobial (AM) films of PHB were prepared by incorporating vanillin (4-hydroxy-3-methoxybenzaldehyde) from 10 to 200 μg/g of PHB. The films were assessed for antimicrobial activity against foodborne pathogens and spoilage bacteria comprising of Escherichia coli, Salmonella typhimurium, Shigella flexneri, and Staphylococcus aureus and fungi such as Aspergillus flavus, Aspergillus fumigatus, Aspergillus niger, Aspergillus parasiticus, Aspergillus ochraceus, Penicillium viridicatum, and Penicillium clavigerum. The minimum concentration of vanillin required to exhibit antimicrobial activity was ≥80 μg/g PHB for bacteria and ≥50 μg/g PHB for fungi. The PHB films with and without vanillin were studied for mechanical and thermal properties such as tensile strength, Young's modulus, percentage elongation to break, melting temperature, and heat of fusion. The thermal stability of the films was studied using thermogravimetric analysis. The release kinetics of vanillin into food matrices was also checked using food stimulants. The study is intended to find applications for PHB films containing vanillin to enhance the shelf life of foods in the form of biodegradable wrapper.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C