Merck
  • Thymoquinone inhibits lipopolysaccharide-induced inflammatory mediators in BV2 microglial cells.
Thymoquinone inhibits lipopolysaccharide-induced inflammatory mediators in BV2 microglial cells.
International immunopharmacology (2015-03-26)
Yanan Wang, Hongmei Gao, Weina Zhang, Wenjie Zhang, Liqun Fang
ZUSAMMENFASSUNG
Thymoquinone, the major active compound isolated from the medicinal Nigella sativa, has been demonstrated to have anti-inflammatory activity. The aim of this study was to investigate the anti-inflammatory effects and mechanisms of thymoquinone on LPS-stimulated BV2 microglial cells. The effects of thymoquinone on inflammatory mediators TNF-α, IL-1β, NO and PGE2 production were detected by ELISA. The effects of thymoquinone on PI3K, Akt phosphorylation, and NF-κB activation were detected by western blot analysis. Our results showed that thymoquinone dose-dependently inhibited LPS-induced TNF-α, IL-1β, NO and PGE2 production. Thymoquinone also inhibited LPS-induced NF-κB activation. Furthermore, thymoquinone was found to inhibit LPS-induced PI3K and Akt phosphorylation, which were upstream molecules of NF-κB. In conclusion, our data demonstrated that thymoquinone might inhibit LPS-induced PI3K and Akt phosphorylation, which leading to the inhibition of NF-κB activation and inflammatory mediator production in BV2 microglia cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymochinon, ≥98%
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic