Merck
Nitric oxide for anammox recovery in a nitrite-inhibited deammonification system.
Environmental technology (2015-04-02)
Ivar Zekker, Ergo Rikmann, Toomas Tenno, Liis Loorits, Kristel Kroon, Hannu Fritze, Tero Tuomivirta, Priit Vabamäe, Markus Raudkivi, Anni Mandel, Sergio S C Dc Rubin, Taavo Tenno
ZUSAMMENFASSUNG
The anaerobic ammonium oxidation (anammox) process is widely used for N-rich wastewater treatment. In the current research the deammonification reactor in a reverse order (first anammox, then the nitrifying biofilm cultivation) was started up with a high maximum N removal rate (1.4 g N m(-2) d(-1)) in a moving bed biofilm reactor. Cultivated biofilm total nitrogen removal rates were accelerated the most by anammox intermediate - nitric oxide (optimum 58 mg NO-N L(-1)) addition. Furthermore, NO was added in order to eliminate inhibition caused by nitrite concentrations (>50 mg [Formula: see text]) increasing [Formula: see text] (2/1, respectively) along with a higher ratio of [Formula: see text] (0.6/1, respectively) than stoichiometrical for this optimal NO amount added during batch tests. Planctomycetales clone P4 sequences, which was the closest (98% and 99% similarity, respectively) relative to Candidatus Brocadia fulgida sequences quantities increase to 1 × 10(6) anammox gene copies g(-1) total suspended solids to till day 650 were determined by quantitative polymerase chain reaction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O