Merck
  • Mannose-centered aromatic galactoclusters inhibit the biofilm formation of Pseudomonas aeruginosa.
Mannose-centered aromatic galactoclusters inhibit the biofilm formation of Pseudomonas aeruginosa.
Organic & biomolecular chemistry (2015-06-20)
Caroline Ligeour, Olivier Vidal, Lucie Dupin, Francesca Casoni, Emilie Gillon, Albert Meyer, Sébastien Vidal, Gérard Vergoten, Jean-Marie Lacroix, Eliane Souteyrand, Anne Imberty, Jean-Jacques Vasseur, Yann Chevolot, François Morvan
ZUSAMMENFASSUNG
Pseudomonas aeruginosa (PA) is a major public health care issue due to its ability to develop antibiotic resistance mainly through adhesion and biofilm formation. Therefore, targeting the bacterial molecular arsenal involved in its adhesion and the formation of its biofilm appears as a promising tool against this pathogen. The galactose-binding LecA (or PA-IL) has been described as one of the PA virulence factors involved in these processes. Herein, the affinity of three tetravalent mannose-centered galactoclusters toward LecA was evaluated with five different bioanalytical methods: HIA, ELLA, SPR, ITC and DNA-based glycoarray. Inhibitory potential towards biofilms was then assessed for the two glycoclusters with highest affinity towards LecA (Kd values of 157 and 194 nM from ITC measurements). An inhibition of biofilm formation of 40% was found for these galactoclusters at 10 μM concentration. Applications of these macromolecules in anti-bacterial therapy are therefore possible through an anti-adhesive strategy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
o-Phenylendiamin, flaked, 99.5%
Sigma-Aldrich
Wasserstoffperoxid-Harnstoffaddukt, 97%
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
o-Phenylendiamin, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylendiamin -dihydrochlorid, peroxidase substrate
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
o-Phenylendiamin, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylendiamin, sublimed, ≥99%
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis