Merck
  • Effect of negatively charged cellulose nanofibers on the dispersion of hydroxyapatite nanoparticles for scaffolds in bone tissue engineering.
Effect of negatively charged cellulose nanofibers on the dispersion of hydroxyapatite nanoparticles for scaffolds in bone tissue engineering.
Colloids and surfaces. B, Biointerfaces (2015-04-26)
Minsung Park, Dajung Lee, Sungchul Shin, Jinho Hyun
ZUSAMMENFASSUNG
Nanofibrous 2,2,6,6-tetramethylpiperidine-1-oxyl(TEMPO)-oxidized bacterial cellulose (TOBC) was used as a dispersant of hydroxyapatite (HA) nanoparticles in aqueous solution. The surfaces of TOBC nanofibers were negatively charged after the reaction with the TEMPO/NaBr/NaClO system at pH 10 and room temperature. HA nanoparticles were simply adsorbed on the TOBC nanofibers (HA-TOBC) and dispersed well in DI water. The well-dispersed HA-TOBC colloidal solution formed a hydrogel after the addition of gelatin, followed by crosslinking with glutaraldehyde (HA-TOBC-Gel). The chemical modification of the fiber surfaces and the colloidal stability of the dispersion solution confirmed TOBC as a promising HA dispersant. Both the Young's modulus and maximum tensile stress increased as the amount of gelatin increased due to the increased crosslinking of gelatin. In addition, the well-dispersed HA produced a denser scaffold structure resulting in the increase of the Young's modulus and maximum tensile stress. The well-developed porous structures of the HA-TOBC-Gel composites were incubated with Calvarial osteoblasts. The HA-TOBC-Gel significantly improved cell proliferation as well as cell differentiation confirming the material as a potential candidate for use in bone tissue engineering scaffolds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
TEMPO, 98%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
Hydroxyapatit, powder, synthetic
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Cetylpyridiniumchlorid, meets USP testing specifications
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
TEMPO, purified by sublimation, 99%
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Calciumphosphat tribasisch, 34.0-40.0% Ca basis
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications