Merck
  • Laboratory Determined Sugar Content and Composition of Commercial Infant Formulas, Baby Foods and Common Grocery Items Targeted to Children.
Laboratory Determined Sugar Content and Composition of Commercial Infant Formulas, Baby Foods and Common Grocery Items Targeted to Children.
Nutrients (2015-07-21)
Ryan W Walker, Michael I Goran
ZUSAMMENFASSUNG
Excess added sugar consumption is tied to poor health outcomes in children. The sugar content of beverages and foods children are exposed to is mostly unknown, yet this information is imperative for understanding potential risks from overconsumption of sugars in early life. We determined actual sugar content by conducting a blinded laboratory analysis in infant formulas, breakfast cereals, packaged baked goods and yogurts. One hundred samples were sent to an independent laboratory for analysis via gas chromatography. Sugar content and composition was determined and total sugar was compared against nutrition labels. Of the 100 samples analyzed, 74% contained ≥20% of total calories per serving from added sugars. Nutrient label data underestimated or overestimated actual sugars and ~25% of all samples had actual total sugar values that were either <10% or >10% of labeled total sugar. Many products that are frequently marketed to and consumed by infants and young children contain sugars in amounts that differ from nutrition labels and often in excess of recommended daily levels. These findings provide further support for adding more comprehensive sugar labeling to food and beverage products, specifically those marketed to, or commonly consumed by, children.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications