Merck
Influence of Staining Method on the Values of Avian Sperm Head Morphometric Variables.
Reproduction in domestic animals = Zuchthygiene (2015-07-21)
S Villaverde-Morcillo, M C Esteso, C Castaño, A Toledano Díaz, A López-Sebastián, J L Campo, J Santiago-Moreno
ZUSAMMENFASSUNG
Computer-assisted systems for the assessment of sperm morphometry (ASMA systems) have been used successfully with several mammalian species. Unfortunately, they have so far been of little use for assessing bird semen, a consequence of the filiform shape of avian spermatozoa. This study compares two staining techniques (Hemacolor(®) and aniline blue staining) for the morphometric analysis of rooster and red-legged partridge spermatozoa as part of a computer-assisted light microscopy method. For both species, Hemacolor(®) staining provided a significantly higher percentage of measurable cells (93.7 ± 11.7% in roosters and 71.9 ± 15.3% in red-legged partridges). Hemacolor(®) also showed greater repeatability (lower coefficients of variation) for length and area in roosters' sperm and for width in the case of red-legged partridge's sperm. In the roosters, the Hemacolor(®) technique returned significantly (p < 0.05) larger sperm head width and area values than did the aniline blue technique, while the latter resulted in greater sperm head length values (p < 0.05). In the red-legged partridge, no differences were seen in the results for sperm head width and area provided by the two techniques, but aniline blue staining was associated with longer length measurements. In conclusion, the morphometric values recorded differed depending on the staining method and species. However, the Hemacolor(®) technique might be deemed the more appropriate for computerized sperm assessment systems as it provides larger percentages of measureable cells and shows greater repeatability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Anilin, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Anilin, ReagentPlus®, 99%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Wasserblau, powder or crystals
Sigma-Aldrich
Wasserblau, certified by the BSC
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C