- Stepwise synthesis and characterization of germa[4], [5], [8], and [10]pericyclynes.
Stepwise synthesis and characterization of germa[4], [5], [8], and [10]pericyclynes.
Dalton transactions (Cambridge, England : 2003) (2015-06-09)
Hiroki Tanimoto, Tomohiko Nagao, Taro Fujiwara, Yasuhiro Nishiyama, Tsumoru Morimoto, Toshimasa Suzuka, Ken Tsutsumi, Kiyomi Kakiuchi
PMID26051845
ZUSAMMENFASSUNG
The stepwise syntheses of germa[N]pericyclynes, including [5]pericyclynes, and their characterization are described. The yields of germa[4] and [8]pericyclynes were improved significantly compared to those obtained in previous studies. The routes reported herein afforded the novel germa[5] and [10]pericyclynes, which were characterized by X-ray crystallography, UV-Vis spectroscopy, and fluorescence emission spectroscopy. A unique fluorescence emission was observed for the large germa[10]pericyclyne ring.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer