Merck
  • Beneficial effects of mineralocorticoid receptor blockade in experimental non-alcoholic steatohepatitis.
Beneficial effects of mineralocorticoid receptor blockade in experimental non-alcoholic steatohepatitis.
Liver international : official journal of the International Association for the Study of the Liver (2015-02-04)
Margarita Pizarro, Nancy Solís, Pablo Quintero, Francisco Barrera, Daniel Cabrera, Pamela Rojas-de Santiago, Juan P Arab, Oslando Padilla, Juan C Roa, Han Moshage, Alexander Wree, Eugenia Inzaugarat, Ariel E Feldstein, Carlos E Fardella, Rene Baudrand, Arnoldo Riquelme, Marco Arrese
ZUSAMMENFASSUNG
Therapeutic options to treat Non-alcoholic steatohepatitis (NASH) are limited. Mineralocorticoid receptor (MR) activation could play a role in hepatic fibrogenesis and its modulation could be beneficial for NASH. To investigate whether eplerenone, a specific MR antagonist, ameliorates liver damage in experimental NASH. C57bl6 mice were fed a choline-deficient and amino acid-defined (CDAA) diet for 22 weeks with or without eplerenone supplementation. Serum levels of aminotransferases and aldosterone were measured and hepatic steatosis, inflammation and fibrosis scored histologically. Hepatic triglyceride content (HTC) and hepatic mRNA levels of pro-inflammatory pro-fibrotic, oxidative stress-associated genes and of MR were also assessed. CDAA diet effectively induced fibrotic NASH, and increased the hepatic expression of pro-inflammatory, pro-fibrotic and oxidative stress-associated genes. Hepatic MR mRNA levels significantly correlated with the expression of pro-inflammatory and pro-fibrotic genes and were significantly increased in hepatic stellate cells obtained from CDAA-fed animals. Eplerenone administration was associated to a reduction in histological steatosis and attenuation of liver fibrosis development, which was associated to a significant decrease in the expression of collagen-α1, collagen type III, alpha 1 and Matrix metalloproteinase-2. The expression of MR correlates with inflammation and fibrosis development in experimental NASH. Specific MR blockade with eplerenone has hepatic anti-steatotic and anti-fibrotic effects. These data identify eplerenone as a potential novel therapy for NASH. Considering its safety and FDA-approved status, human studies are warranted.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aldosteron, ≥95% (HPLC)
Sigma-Aldrich
DL-Alanin, ≥99% (HPLC)
Sigma-Aldrich
DL-Alanin, ≥99%, FCC, FG
Sigma-Aldrich
Eplerenon, ≥98% (HPLC)