Merck
  • Preparation and Characterization of Composites Based on Polylactic Acid and Beeswax with Improved Water Vapor Barrier Properties.
Preparation and Characterization of Composites Based on Polylactic Acid and Beeswax with Improved Water Vapor Barrier Properties.
Journal of food science (2015-10-08)
Jung Hoon Lim, Jeong Ae Kim, Jung A Ko, Hyun Jin Park
ZUSAMMENFASSUNG
Beeswax and a plasticizer (ATBC) were added to polylactic acid (PLA) films in order to enhance the water vapor barrier properties of the films. Beeswax improved the barrier properties; the water vapor permeability in the composite containing 1% beeswax was 58% lower than that of the neat PLA. Fourier transform infrared spectroscopy and X-ray diffraction analysis revealed that the incorporation of beeswax and ATBC had so little effect on the PLA structure. In addition, the structure of PLA did not vary substantially with the additions. The surfaces of the composites were examined by using field emission scanning electron microscopy. Differential scanning calorimetry results showed that the degree of crystallinity of the PLA films increased with the addition of beeswax and ATBC. However, the tensile strength and elongation at break of the composites containing beeswax were up to approximately 50% lower than those of the neat PLA. Although further study is needed to improve the mechanical properties, the aforementioned results showed that the PLA barrier properties can be improved by the incorporation of a small amount of beeswax and ATBC. The results of this study can be applied for the preparation of PLA composite films with improved barrier properties. Such biodegradable films are extremely useful for applications in the food packaging industry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tributyl-O-acetylcitrat, 98%