Merck
  • Biocompatibility of MgF2-coated MgNd2 specimens in contact with mucosa of the nasal sinus - a long term study.
Biocompatibility of MgF2-coated MgNd2 specimens in contact with mucosa of the nasal sinus - a long term study.
Acta biomaterialia (2015-03-15)
Constantin M Weber, Rainer Eifler, Jan-Marten Seitz, Hans J Maier, Janin Reifenrath, Thomas Lenarz, Martin Durisin
ZUSAMMENFASSUNG
Up to now, different surgical techniques and stent systems have already been developed and tested for the continuous and adequate ventilation of the frontal sinuses. However, the results achieved still remain poor. Magnesium-based implants have been successfully used in numerous clinical applications. Offering excellent biocompatibility and biodegradability it may be the ideal material for the development of novel implants of the nasal sinus. Here, we present for the first time results on the behaviour of magnesium alloy in a unique environment, i.e. in contact to the nasal mucosa, air and nasal secretion. In a prospective longitudinal study, magnesium fluoride-coated MgNd2 specimens were implanted in the frontal sinuses of 12 minipigs for the investigation of biocompatibility and of the interface between the implant and the mucosa. Endoscopic examinations, histopathological evaluation and EDX measurements were performed regularly up to 180days. Endoscopic evaluation showed focal mucosal reaction, however, without affecting the patency of the sinus. In addition, no signs of bacterial infections were observed. The EDX analyses showed a marginal but steady increase in the Mg concentration in the mucosa over 180days. Histological analysis revealed a locally confined moderate mucosal hyperplasia and unspecific inflammatory reaction. Furthermore, we did not find any osteoinductive effects of the magnesium alloy. The results indicate the excellent biocompatibility of the MgNd2 alloy in contact with nasal mucosa and provide a novel material compound and solid proof-of-principle for the development of magnesium-based nasal stents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Neodym, powder, −40 mesh, ≥99% trace rare earth metals basis
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium Zubereitung, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium (in Sure/Seal Flasche), in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Sigma-Aldrich
Magnesium (in Sure/Seal Flasche), in a Sure/Seal bottle, turnings, 37.5 mmol
Neodym, rod, 50mm, diameter 12.5mm, cast, 99%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%