Merck
Thiolate versus Selenolate: Structure, Stability, and Charge Transfer Properties.
ACS nano (2015-04-11)
Jakub Ossowski, Tobias Wächter, Laura Silies, Martin Kind, Agnieszka Noworolska, Florian Blobner, Dominika Gnatek, Jakub Rysz, Michael Bolte, Peter Feulner, Andreas Terfort, Piotr Cyganik, Michael Zharnikov
ZUSAMMENFASSUNG
Selenolate is considered as an alternative to thiolate to serve as a headgroup mediating the formation of self-assembled monolayers (SAMs) on coinage metal substrates. There are, however, ongoing vivid discussions regarding the advantages and disadvantages of these anchor groups, regarding, in particular, the energetics of the headgroup-substrate interface and their efficiency in terms of charge transport/transfer. Here we introduce a well-defined model system of 6-cyanonaphthalene-2-thiolate and -selenolate SAMs on Au(111) to resolve these controversies. The exact structural arrangements in both types of SAMs are somewhat different, suggesting a better SAM-building ability in the case of selenolates. At the same time, both types of SAMs have similar packing densities and molecular orientations. This permitted reliable competitive exchange and ion-beam-induced desorption experiments which provided unequivocal evidence for a stronger bonding of selenolates to the substrate as compared to the thiolates. Regardless of this difference, the dynamic charge transfer properties of the thiolate- and selenolate-based adsorbates were found to be nearly identical, as determined by the core-hole-clock approach, which is explained by a redistribution of electron density along the molecular framework, compensating the difference in the substrate-headgroup bond strength.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäureanhydrid, ReagentPlus®, ≥99%