Merck
Ultrafine Nanoparticle-Supported Ru Nanoclusters with Ultrahigh Catalytic Activity.
Small (Weinheim an der Bergstrasse, Germany) (2015-06-18)
Lihua Zhu, Yingying Jiang, Jinbao Zheng, Nuowei Zhang, Changlin Yu, Yunhua Li, Chih-Wen Pao, Jeng-Lung Chen, Chuanhong Jin, Jyh-Fu Lee, Chuan-Jian Zhong, Bing H Chen
ZUSAMMENFASSUNG
The design of an ideal heterogeneous catalyst for hydrogenation reaction is to impart the catalyst with synergetic surface sites active cooperatively toward different reaction species. Herein a new strategy is presented for the creation of such a catalyst with dual active sites by decorating metal and metal oxide nanoparticles with ultrafine nanoclusters at atomic level. This strategy is exemplified by the design and synthesis of Ru nanoclusters supported on Ni/NiO nanoparticles. This Ru-nanocluster/Ni/NiO-nanoparticle catalyst is shown to exhibit ultrahigh catalytic activity for benzene hydrogenation reaction, which is 55 times higher than Ru-Ni alloy or Ru on Ni catalysts. The nanoclusters-on-nanoparticles are characterized by high-resolution transmission electron microscope, Cs-corrected high angle annular dark field-scanning transmission electron microscopy, elemental mapping, high-sensitivity low-energy ion scattering, and X-ray absorption spectra. The atomic-scale nanocluster-nanoparticle structural characteristics constitute the basis for creating the catalytic synergy of the surface sites, where Ru provides hydrogen adsorption and dissociation site, Ni acts as a "bridge" for transferring H species to benzene adsorbed and activated at NiO site, which has significant implications to multifunctional nanocatalysts design for wide ranges of catalytic reactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Hydrazin Monohydrat, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazin Hydrat, reagent grade, N2H4 50-60 %
Sigma-Aldrich
2-Pyrrolidinon, ≥99%
Sigma-Aldrich
2-Pyrrolidinon, 99%
Sigma-Aldrich
Cyclohexan, anhydrous, 99.5%
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure, technical grade
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O