Merck
  • Ibuprofen lysinate, quicker and less variable: relative bioavailability compared to ibuprofen base in a pediatric suspension dosage form.
Ibuprofen lysinate, quicker and less variable: relative bioavailability compared to ibuprofen base in a pediatric suspension dosage form.
International journal of clinical pharmacology and therapeutics (2015-08-08)
Juan Manuel Ferrero-Cafiero, Ignasi Gich, Montse Puntes, Joan Martínez, Maria R Ballester, Jimena Coimbra, Yaira Mathison, Maite Tarré, Xavier Font, Rosa M Antonijoan
ZUSAMMENFASSUNG
To assess and compare the bioavailability of ibuprofen enantiomers (R and S) of two different pediatric suspensions: the first one with ibuprofen lysinate (Algidrin® Pediátrico, FARDI S.A., Barcelona, Spain) and the second one with ibuprofen base (Dalsy®, Abbott Laboratories S.A., Madrid, Spain). A randomized, open-label, single-dose, balanced, crossover study under fasting conditions was performed at the CIM-Sant Pau. 24 healthy volunteers received a single dose of ibuprofen lysinate (Algidrin® Pediátrico, FARDI S.A.) and ibuprofen base (Dalsy®, Abbott Laboratories S.A.) equivalent to 400 mg of ibuprofen. 18 blood samples were drawn and ibuprofen enantiomer plasma concentrations were determined using an enantioselective analytical method. An analysis of variance (ANOVA) model was used, and the 90% confidence intervals (CI) were calculated; further analyses were made regarding rate of absorption and variability. The pharmacokinetic parameters (Algidrin® Pediátrico vs. Dalsy® (Mean±SD)) were: S-enantiomer: Cmax=22.39±5.33 vs. 19.97±3.19 μg/mL; AUC0t=74.83±16.69 vs. 74.64±14.80 μg×h/mL, and AUC0∞=77.46±19.33 vs. 76.98±17.13 μg×h/mL; and for R-enantiomer: Cmax=21.74±3.76 vs. 15.20±2.03 μg/mL; AUC0t=57.55±10.17 vs. 46.13±9.61 μg×h/mL, and AUC0∞ value was 58.49±10.57 vs. 47.03±10.02 μg×h/mL. The tmax (Median) for S-enantiomer (active) were: 0.5 vs. 1.33 hours (p=0.001) and for R-enantiomer: 0.5 vs. 1.0 hours (p=0.004). Ibuprofen pharmacokinetic values may vary under fed state and in pediatric population. While S-ibuprofen shows a similar bioavailability for AUC0t, AUC0∞, and Cmax, R-ibuprofen shows suprabioavailability for the lysinate formulation. The rate of absorption of the ibuprofen lysinate suspension is quicker and less variable than that of the ibuprofen base reference suspension and it exhibits a shorter tmax, which is of particular interest for achieving a rapid and homogeneous analgesic and antipyretic effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications