Merck
  • Preliminary in vitro and ex vivo evaluation of afzelin, kaempferitrin and pterogynoside action over free radicals and reactive oxygen species.
Preliminary in vitro and ex vivo evaluation of afzelin, kaempferitrin and pterogynoside action over free radicals and reactive oxygen species.
Archives of pharmacal research (2014-10-16)
José Carlos Rebuglio Vellosa, Luis Octávio Regasini, Caroline Belló, Josiane Aparecida Schemberger, Najeh Maissar Khalil, Andreia de Araújo Morandim-Giannetti, Vanderlan da Silva Bolzani, Iguatemy Lourenço Brunetti, Olga Maria Mascarenhas de Faria Oliveira
ZUSAMMENFASSUNG
Biological activities of flavonoids have been extensively reviewed in literature. The biochemical profile of afzelin, kaempferitrin, and pterogynoside acting on reactive oxygen species was investigated in this paper. The flavonoids were able to act as scavengers of the superoxide anion, hypochlorous acid and taurine chloramine. Although flavonoids are naturally occurring substances in plants which antioxidant activities have been widely advertised as beneficial, afzelin, kaempferitrin, and pterogynoside were able to promote cytotoxic effect. In red blood cells this toxicity was enhanced, depending on flavonoids concentration, in the presence of hypochlorous acid, but reduced in the presence of 2,2'-azo-bis(2-amidinopropane) free radical. These flavonoids had also promoted the death of neutrophils, which was exacerbated when the oxidative burst was initiated by phorbol miristate acetate. Therefore, despite their well-known scavenging action toward free radicals and oxidants, these compounds could be very harmful to living organisms through their action over erythrocytes and neutrophils.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Adenin, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥99%
Sigma-Aldrich
Phenazin-Methosulfat
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, ≥90.0% (HPLC)
Sigma-Aldrich
Adenin, BioReagent, suitable for cell culture
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98% (TLC)
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98.0% (NT)
Sigma-Aldrich
Nitrotetrazolium-Blauchlorid, powder, electrophoresis grade
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Adenin, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Kaempferitrin, ≥97% (NMR)