Merck
  • Fluxes in PHA-storing microbial communities during enrichment and biopolymer accumulation processes.
Fluxes in PHA-storing microbial communities during enrichment and biopolymer accumulation processes.
New biotechnology (2015-08-11)
Om Murugan Janarthanan, Bronwyn Laycock, Liliana Montano-Herrera, Yang Lu, Monica V Arcos-Hernandez, Alan Werker, Steven Pratt
ZUSAMMENFASSUNG
The use of mixed microbial cultures for the production of polyhydroxyalkanoates (PHAs) is emerging as a viable technology. In this study, 16S rRNA gene amplicon pyrosequencing was used to analyse fluctuations in populations over a 63-day period within a PHA-storing mixed microbial community enriched on fermented whey permeate. This community was dominated by the genera Flavisolibacter and Zoogloea as well as an unidentified organism belonging to the phylum Bacteroidetes. The population was observed to cycle through an increase in Zoogloea followed by a return to a community composition similar to the initial one (highly enriched in Flavisolibacter). It was found that the PHA accumulation capacity of the community was robust to population flux during enrichment and even PHA accumulation, with final polymer composition dependent on the overall proportion of acetic to propionic acids in the feed. This community adaptation suggests that mixed culture PHA production is a robust process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Phosphonohexansäure, 97%