Merck
  • Formaldehyde migration in aqueous extracts from paper and cardboard food packaging materials in Turkey.
Formaldehyde migration in aqueous extracts from paper and cardboard food packaging materials in Turkey.
Food additives & contaminants. Part B, Surveillance (2015-06-23)
Canan Ekinci Dogan, Rukiye Sancı
ZUSAMMENFASSUNG
Migration of formaldehyde to aqueous extracts from paper and cardboard food packaging materials was determined by an ultraviolet visible-spectrophotometric method at 410 nm. Intraday and interday precision of the method, expressed as coefficient of variation, varied between 1.5 to 4.4% and 7 to 8.8%, respectively. The limit of quantification was 0.28 mg kg(-1) for formaldehyde in aqueous extracts. The recovery of the method was over 90% for two different concentration levels in aqueous extracts. The method was applied to the migration of formaldehyde to aqueous extracts from 31 different paper and cardboard materials collected from the packaging sector, intended for food contact, such as tea filters, hot water filters, paper pouches and folding boxes. The results were between limit of detection 0.23 mg/kg and 40 mg kg(-1) and were evaluated according to the relevant directives.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Acetylaceton, Wacker Chemie AG, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C