Merck
  • Presence of delta-(L-alpha-aminoadipyl)-L-cysteinyl-D-valine in fermentations of Penicillium chrysogenum.
Presence of delta-(L-alpha-aminoadipyl)-L-cysteinyl-D-valine in fermentations of Penicillium chrysogenum.
Antimicrobial agents and chemotherapy (1975-12-01)
P Adriaens, B Meesschaert, W Wuyts, H Vanderhaeghe, H Eyssen
PMID813571
ZUSAMMENFASSUNG
Cultures of Penicillium chrysogenum, growth with [(35)S]sulfate or labeled amino acids, were examined by ion-exchange chromatography for possible peptidic precursors of penicillin. A sulfur-containing compound, present in both the mycelial extracts and the culture filtrates, was eluted at the location of the synthetic lld-tripeptide delta-(l-alpha-aminoadipyl)-l-cysteinyl-d-valine. Since this compound was also labeled when the cultures were incubated with dl-[6-(14)C]alpha-aminoadipic acid, l-[3,3'-(3)H]cystine, or dl-[1-(14)C]valine, its identity with the synthetic lld-tripeptide can be accepted. No delta-(l-alpha-aminoadipyl)-l-cysteine or lll-tripeptide were detected. The implications of these findings for tripeptide and penicillin biosynthesis are discussed.