Merck
  • Gene electrotransfer of plasmid AMEP, an integrin-targeted therapy, has antitumor and antiangiogenic action in murine B16 melanoma.
Gene electrotransfer of plasmid AMEP, an integrin-targeted therapy, has antitumor and antiangiogenic action in murine B16 melanoma.
Gene therapy (2015-03-18)
M Bosnjak, T Dolinsek, M Cemazar, S Kranjc, T Blagus, B Markelc, M Stimac, J Zavrsnik, U Kamensek, L Heller, C Bouquet, B Turk, G Sersa
ZUSAMMENFASSUNG
Gene therapy with Plasmid AMEP (antiangiogenic metargidin peptide) has recently been studied as a potential targeted therapy for melanoma. This plasmid is designed to downregulate α5β1 and αvβ3 integrins. In our study, electroporation was used as a nonviral delivery system. We investigated the antiangiogenic and direct antitumor effectiveness of this gene therapy on low and highly metastatic B16 melanoma variants. In vitro, the antiangiogenic effectiveness as determined by tube formation assay on endothelial cells was predominantly dependent on AMEP expression levels. In vivo, antitumor effectiveness was mediated by the inhibition of proliferation, migration and invasion of melanoma cells and correlated with the expression of integrins on tumor cells after intratumor delivery. In addition, reduced metastatic potential was shown. Intramuscular gene electrotransfer of Plasmid AMEP, for AMEP systemic distribution, had no antitumor effect with this specific preventive treatment protocol, confirming that direct tumor delivery was more effective. This study confirms our previous in vitro data that the expression levels of integrins on melanoma cells could be used as a biomarker for antitumor effectiveness in integrin-targeted therapies, whereas the expression levels of AMEP peptide could be a predictive factor for antiangiogenic effectiveness of Plasmid AMEP in the treatment of melanoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Rhodamin B, ≥95% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Rhodamin B, suitable for fluorescence
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Calcein-AM -Lösung, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Rhodamin B, Dye content 90 %