Merck
Comparison of milk oligosaccharides pattern in colostrum of different horse breeds.
Journal of agricultural and food chemistry (2015-05-01)
E Difilippo, H A M Willems, J C Vendrig, J Fink-Gremmels, H Gruppen, H A Schols
ZUSAMMENFASSUNG
Colostrum oligosaccharides are known to exhibit prebiotic and immunomodulatory properties. Oligosaccharide composition is species-specific, and equine colostrum has been reported to contain unique oligosaccharides. Therefore, equine oligosaccharides (EMOS) from colostrum from different horse breeds were analyzed by CE-LIF, CE-MS(n), HILIC-MS(n), and exoglycosidase degradation. Sixteen EMOS were characterized and quantified, of which half were neutral and half were acidic. EMOS showed about 63% structural overlap with human milk oligosaccharides, known for their bioactivity. Seven EMOS were not reported before in equine oligosaccharides literature: neutral Gal(β1-4)HexNAc, Gal(β1-4)Hex-Hex, β4'-galactosyllactose, and lactose-N-hexaose, as well as acidic 6'-Sialyl-Hex-Ac-HexNAc, sialyllacto-N-tetraose-a, and disialylacto-N-tetraose (isomer not further specified). In all colostrum samples, the average oligosaccharide concentration ranged from 2.12 to 4.63 g/L; with β 6'and 3'- galactosyllactose, 3'-sialyllactose, and disialyllactose as the most abundant of all oligosaccharides (27-59, 16-37, 1-8, and 1-6%, respectively). Differences in presence and in abundance of specific EMOS were evident not only between the four breeds but also within the breed.