Merck
  • Preparation and evaluation of monodispersed, submicron, non-porous silica particles functionalized with β-CD derivatives for chiral-pressurized capillary electrochromatography.
Preparation and evaluation of monodispersed, submicron, non-porous silica particles functionalized with β-CD derivatives for chiral-pressurized capillary electrochromatography.
Electrophoresis (2015-05-21)
Lu Yangfang, Wang Hui, Xue Yun, Gu Xue, Wang Yan, Yan Chao
ZUSAMMENFASSUNG
Submicron, non-porous, chiral silica stationary phase has been prepared by the immobilization of functionalized β-CD derivatives to isocyanate-modified silica via chemical reaction and applied to the pressurized capillary electrochromatography (pCEC) enantio-separation of various chiral compounds. The submicron, non-porous, cyclodextrin-based chiral stationary phases (sub_μm-CSP2) exhibited excellent chiral recognition of a wide range of analytes including clenbuterol hydrochloride, mexiletine hydrochloride, chlorpheniramine maleate, esmolol hydrochloride, and metoprolol tartrate. The synthesized submicron particles were regularly spherical and uniformly non-porous with an average diameter of around 800 nm and a mean pore size of less than 2 nm. The synthesized chiral stationary phase was packed into 10 cm × 100 μm id capillary columns. The sub_μm-CSP2 column used in the pCEC system showed better separation of the racemates and at a higher rate compared to those used in the capillary liquid chromatography mode (cLC) system. The sub_μm-CSP2 possessed high mechanical strength, high stereoselectivity, and long lifespan, demonstrating rapid enantio-separation and good resolution of samples. The column provided an efficiency of up to 170,000 plates/m for n-propylbenzene.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Tetraethylorthosilikat, 99.999% trace metals basis
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure, technical grade
Sigma-Aldrich
Tetraethylorthosilicat, packaged for use in deposition systems
Sigma-Aldrich
4,5-Dihydroxy-anthrachinon-2-carbonsäure