Merck
  • Quantification of Change in Pigment Epithelial Detachment Volume and Morphology After Transition to Intravitreal Aflibercept in Eyes With Recalcitrant Neovascular AMD: 18-Month Results.
Quantification of Change in Pigment Epithelial Detachment Volume and Morphology After Transition to Intravitreal Aflibercept in Eyes With Recalcitrant Neovascular AMD: 18-Month Results.
Ophthalmic surgery, lasers & imaging retina (2015-06-27)
Aditya Kanesa-Thasan, Dilraj S Grewal, Manjot K Gill, Alice T Lyon, Rukhsana G Mirza
ZUSAMMENFASSUNG
To quantitatively evaluate the change in pigment epithelial detachment (PED) morphology on spectral-domain optical coherence tomography (SD-OCT) 18 months after the transition to intravitreal aflibercept in patients with neovascular age-related macular degeneration (AMD) with PED recalcitrant to monthly intravitreal bevacizumab or ranibizumab. Retrospective case series examining patients with neovascular AMD who had a persistent fibrovascular or serous PED on SD-OCT. PED volume was calculated by manually outlining the PED on individual OCT slices of the raster scan and multiplying by the pixel dimensions. Eleven eyes of 10 patients who had received an average of 25.7 ± 20.1 (range: 6 to 70) prior bevacizumab or ranibizumab injections over a period of 26.6 ± 19.8 months (range: 4 to 63) were included. PED volume decreased with aflibercept from 0.687 ± 0.837 mm(3) to 0.562 ± 0.705 mm(3) (P = .02), a decrease of 19% ± 12.27%. After 18 months of aflibercept, recalcitrant PED volumes were reduced by 19% while preserving visual acuity in eyes with neovascular AMD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Di(N-succinimidyl)-Glutarat, ≥97.0% (CHN)
Sigma-Aldrich
Triamcinolon