Merck
Turning Squalene into Cationic Lipid Allows a Delivery of siRNA in Cultured Cells.
Nucleic acid therapeutics (2015-04-22)
Jean-Rémi Bertrand, Claire Lucas, Ngoc Minh Pham, Catherine Durieu, Patrick Couvreur, Claude Paul Malvy, Didier Desmaële
ZUSAMMENFASSUNG
Covalent binding of squalene to siRNA has already been shown to be an interesting way of delivering siRNA in vivo. Whether squalene derivatives could also be used to deliver siRNA in cells without covalent binding similar to usual transfection with cationic lipids is the question addressed in this article. Accordingly, we investigated the activity of two squalene derivatives bearing a quaternary ammonium head group and a guanidinium group, respectively. The second derivative displayed interesting properties for delivering siRNA into cells in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Squalen, ≥98%, liquid
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Isoamylalkohol, ≥98%, FG
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trimethylamin -hydrochlorid, 98%
Sigma-Aldrich
3-Methyl-1-Butanol, anhydrous, ≥99%
Sigma-Aldrich
Aminoguanidin-hydrogencarbonat, 97%
Sigma-Aldrich
Isoamylalkohol, natural, ≥98%, FG
Sigma-Aldrich
3-Methylbutanol, BioReagent, ≥98.5%, Molecular Biology
Sigma-Aldrich
3-Methylbutanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
MISSION® esiRNA, targeting human FLI1