Merck
  • Application of enzymatic apple pomace hydrolysate to production of 2,3-butanediol by alkaliphilic Bacillus licheniformis NCIMB 8059.
Application of enzymatic apple pomace hydrolysate to production of 2,3-butanediol by alkaliphilic Bacillus licheniformis NCIMB 8059.
Journal of industrial microbiology & biotechnology (2015-10-09)
Aneta M Białkowska, Ewa Gromek, Joanna Krysiak, Barbara Sikora, Halina Kalinowska, Marzena Jędrzejczak-Krzepkowska, Celina Kubik, Siegmund Lang, Fokko Schütt, Marianna Turkiewicz
ZUSAMMENFASSUNG
2,3-Butanediol (2,3-BD) synthesis by a nonpathogenic bacterium Bacillus licheniformis NCIMB 8059 from enzymatic hydrolysate of depectinized apple pomace and its blend with glucose was studied. In shake flasks, the maximum diol concentration in fed-batch fermentations was 113 g/L (in 163 h, from the hydrolysate, feedings with glucose) while in batch processes it was around 27 g/L (in 32 h, from the hydrolysate and glucose blend). Fed-batch fermentations in the 0.75 and 30 L fermenters yielded 87.71 g/L 2,3-BD in 160 h, and 72.39 g/L 2,3-BD in 94 h, respectively (from the hydrolysate and glucose blend, feedings with glucose). The hydrolysate of apple pomace, which was for the first time used for microbial 2,3-BD production is not only a source of sugars but also essential minerals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salpetersäure, ACS reagent, 70%
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Salpetersäure, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Salpetersäure, red, fuming, HNO3 >90 %
Sigma-Aldrich
trans-Ferulasäure, 99%
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Sigma-Aldrich
3-Hydroxy-benzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
trans-Ferulasäure, ≥99%
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N