- Isolation of High-Purity Extracellular Vesicles by Extracting Proteins Using Aqueous Two-Phase System.
Isolation of High-Purity Extracellular Vesicles by Extracting Proteins Using Aqueous Two-Phase System.
PloS one (2015-06-20)
Jongmin Kim, Hyunwoo Shin, Jiyoon Kim, Junho Kim, Jaesung Park
PMID26090684
ZUSAMMENFASSUNG
We present a simple and rapid method to isolate extracellular vesicles (EVs) by using a polyethylene glycol/dextran aqueous two-phase system (ATPS). This system isolated more than ~75% of melanoma-derived EVs from a mixture of EVs and serum proteins. To increase the purity of EVs, a batch procedure was combined as additional steps to remove protein contaminants, and removed more than ~95% of the protein contaminants. We also performed RT-PCR and western blotting to verify the diagnostic applicability of the isolated EVs, and detected mRNA derived from melanoma cells and CD81 in isolated EVs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles