Merck
  • Arteriosclerosis in zero-time biopsy is a risk factor for tacrolimus-induced chronic nephrotoxicity.
Arteriosclerosis in zero-time biopsy is a risk factor for tacrolimus-induced chronic nephrotoxicity.
Nephrology (Carlton, Vic.) (2015-06-03)
Takafumi Yagisawa, Kazuya Omoto, Tomokazu Shimizu, Hideki Ishida, Kazunari Tanabe
ZUSAMMENFASSUNG
Calcineurin inhibitors reduce the acute rejection rate and greatly improve renal allograft survival. However, they are associated with some adverse events, including nephrotoxicity, a risk factor for allograft failure. Chronic calcineurin inhibitor-induced nephrotoxicity causes irreversible damage to renal components, such as arteriolar hyaline thickening. The aim of this study is to investigate the risk factors for tacrolimus-induced chronic nephrotoxicity using zero-time biopsy specimens. Between January 2001 and December 2010, 483 patients who underwent living-related kidney transplantation and had also been placed on a tacrolimus-based regimen were enrolled in this study. There were 1859 specimens evaluated comprising 483 zero-time biopsy specimens and 1376 protocol and for-cause biopsy specimens. De novo arteriolar hyaline thickening due to tacrolimus-induced chronic nephrotoxicity was scored according to the Banff classification aah score. In this study, tacrolimus-induced nephrotoxicity was defined as a positive aah score. Of the 483 patients, 108 patients (22.4%) had biopsy-proven tacrolimus-induced chronic nephrotoxicity. Multivariate analysis showed that interlobular arteriosclerosis proven by zero-time biopsy (OR: 2.23, 95%CI: 1.38-3.58, P < 0.01) and acute rejection episodes (OR: 1.58, 95%CI: 1.00-2.47, P = 0.04) were independent risk factors for tacrolimus-induced chronic nephrotoxicity. However, tacrolimus-induced chronic nephrotoxicity did not affect long-term graft survival. This is the first report showing that arteriosclerosis in zero-time biopsy specimens is a risk factor for histological tacrolimus-induced chronic nephrotoxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
FK-506 Monohydrat, ≥98% (HPLC)
Sigma-Aldrich
6α-Methylprednisolon, ≥98%
Sigma-Aldrich
Mycophenolat-Mofetil, ≥98% (HPLC)