Merck
Reversal of P-gp and BCRP-mediated MDR by tariquidar derivatives.
European journal of medicinal chemistry (2015-07-22)
Xu-Qin Li, Lin Wang, Yan Lei, Tao Hu, Fei-Long Zhang, Chi-Hin Cho, Kenneth K W To
ZUSAMMENFASSUNG
With an aim to generate non-toxic, specific and highly potent multidrug resistance (MDR) modulators, a novel series of anthranilic acid amide-substituted tariquidar derivatives were synthesized. The new compounds were evaluated for their cytotoxicity toward normal human colon fibroblasts (CCD18-Co), human gastric epithelial cell line (HFE) and primary rat liver cells, and for their ability to inhibit P-gp/BCRP-mediated drug efflux and reversal of P-gp and BCRP-mediated MDR in parental and drug-resistant cancer cell lines (LCC6 MDR1, MCF-7 FLV1000, R-HepG2, SW620-Ad300). While tariquidar is highly toxic to normal cells, the new derivatives exhibited much lower or negligible cytotoxicity. Some of the new tariquidar derivatives inhibited both P-gp and BCRP-mediated drug efflux whereas a few of them bearing a sulfonamide functional group (1, 5, and 16) are specific to P-gp. The new compounds were also found to potentiate the anticancer activity of the transporter substrate anticancer drugs in the corresponding transporter-overexpressing cell lines. The extent of resistance reversal was found to be consistent with the transporter inhibitory effect of the new derivatives. To further understand the mechanism of P-gp and BCRP inhibition, the tariquidar derivatives were found to interact with the transporters using an antibody-based UIC2 or 5D3 shift assay. Moreover, the transporters-inhibiting derivatives were found to modulate the ATPase activities of the two MDR transporters. Our data thus advocate further development of the new compounds for the circumvention of MDR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Doxorubicin -hydrochlorid, 98.0-102.0% (HPLC)
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Doxorubicin -hydrochlorid, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminiumoxid, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminiumoxid, 99.997% trace metals basis
Sigma-Aldrich
Aluminiumoxid, single crystal substrate, <0001>
Sigma-Aldrich
Aluminiumoxid, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminumoxid, mesoporös, MSU-X (wormhole), average pore size 3.8 nm