Merck
  • The Cyclooctadepsipeptide Anthelmintic Emodepside Differentially Modulates Nematode, Insect and Human Calcium-Activated Potassium (SLO) Channel Alpha Subunits.
The Cyclooctadepsipeptide Anthelmintic Emodepside Differentially Modulates Nematode, Insect and Human Calcium-Activated Potassium (SLO) Channel Alpha Subunits.
PLoS neglected tropical diseases (2015-10-06)
Anna Crisford, Ulrich Ebbinghaus-Kintscher, Eva Schoenhense, Achim Harder, Klaus Raming, Ita O'Kelly, Kelechi Ndukwe, Vincent O'Connor, Robert J Walker, Lindy Holden-Dye
ZUSAMMENFASSUNG
The anthelmintic emodepside paralyses adult filarial worms, via a mode of action distinct from previous anthelmintics and has recently garnered interest as a new treatment for onchocerciasis. Whole organism data suggest its anthelmintic action is underpinned by a selective activation of the nematode isoform of an evolutionary conserved Ca2+-activated K+ channel, SLO-1. To test this at the molecular level we compared the actions of emodepside at heterologously expressed SLO-1 alpha subunit orthologues from nematode (Caenorhabditis elegans), Drosophila melanogaster and human using whole cell voltage clamp. Intriguingly we found that emodepside modulated nematode (Ce slo-1), insect (Drosophila, Dm slo) and human (hum kcnma1)SLO channels but that there are discrete differences in the features of the modulation that are consistent with its anthelmintic efficacy. Nematode SLO-1 currents required 100 μM intracellular Ca2+ and were strongly facilitated by emodepside (100 nM; +73.0 ± 17.4%; n = 9; p < 0.001). Drosophila Slo currents on the other hand were activated by emodepside (10 μM) in the presence of 52 nM Ca2+ but were inhibited in the presence of 290 nM Ca2+ and exhibited a characteristic loss of rectification. Human Slo required 300 nM Ca2+ and emodepside transiently facilitated currents (100 nM; +33.5 ± 9%; n = 8; p<0.05) followed by a sustained inhibition (-52.6 ± 9.8%; n = 8; p < 0.001). This first cross phyla comparison of the actions of emodepside at nematode, insect and human channels provides new mechanistic insight into the compound's complex modulation of SLO channels. Consistent with whole organism behavioural studies on C. elegans, it indicates its anthelmintic action derives from a strong activation of SLO current, not observed in the human channel. These data provide an important benchmark for the wider deployment of emodepside as an anthelmintic treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Kalzium-Ionophor A23187, ≥98% (TLC), powder
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Kaliumhydroxid, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Sigma-Aldrich
Kaliumhydroxid, technical, ≥85%, powder
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Kaliumhydroxid, ≥99.95% trace metals basis
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis