Merck
  • Clearance of Pneumococcal Colonization in Infants Is Delayed through Altered Macrophage Trafficking.
Clearance of Pneumococcal Colonization in Infants Is Delayed through Altered Macrophage Trafficking.
PLoS pathogens (2015-06-25)
Steven J Siegel, Edwin Tamashiro, Jeffrey N Weiser
ZUSAMMENFASSUNG
Infections are a common cause of infant mortality worldwide, especially due to Streptococcus pneumoniae. Colonization is the prerequisite to invasive pneumococcal disease, and is particularly frequent and prolonged in children, though the mechanisms underlying this susceptibility are unknown. We find that infant mice exhibit prolonged pneumococcal carriage, and are delayed in recruiting macrophages, the effector cells of clearance, into the nasopharyngeal lumen. This lack of macrophage recruitment is paralleled by a failure to upregulate chemokine (C-C) motif ligand 2 (Ccl2 or Mcp-1), a macrophage chemoattractant that is required in adult mice to promote clearance. Baseline expression of Ccl2 and the related chemokine Ccl7 is higher in the infant compared to the adult upper respiratory tract, and this effect requires the infant microbiota. These results demonstrate that signals governing macrophage recruitment are altered at baseline in infant mice, which prevents the development of appropriate innate cell infiltration in response to pneumococcal colonization, delaying clearance of pneumococcal carriage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Metronidazol, BioXtra
Sigma-Aldrich
Ampicillin, meets USP testing specifications