Merck
  • Calcium-ion-modulated ceramic hydroxyapatite resin for the scalable purification of recombinant Adeno-Associated Virus serotype 9.
Calcium-ion-modulated ceramic hydroxyapatite resin for the scalable purification of recombinant Adeno-Associated Virus serotype 9.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-04-05)
Weihong Qu, Mingxi Wang, Yaqing Wu, Yinghui Lv, Qizhao Wang, Ruian Xu
ZUSAMMENFASSUNG
Column chromatography has been widely used as a scalable purification strategy for recombinant adeno-associated virus (rAAV) vectors. The rAAV1, 2, 4, 5, 6, 8 and 9 serotypes could be separated using affinity resins, ion exchange resins or other types of resins. Apatite resin has displayed outstanding performance in protein purification in the past 10 years, and ceramic hydroxyapatite (CHT) chromatography resin with a polyethylene glycol (PEG) modulation has recently been used for rAAV1 and rAAV9 vectors. This study reports the use of CHT chromatography modulated by calcium ions instead of PEG for rAAV9 purification. Calcium-ion-containing buffers effectively improve the inclusion of CHT as a capture resin, the resin-binding capacity and the yield. The optimum calcium ion concentration is 30ppm, and the optimum pH is 7.0. A frontal analysis indicated that the binding capacity of CHT at 2ml/min reaches 65.1mg total protein per ml of resin. A previously developed purification strategy consists of CHT followed by ANX anion exchange chromatography. The vector yield of this approach is approximately 70%, and a software analysis indicated a vector purity exceeding 98%. The residual host cell (HEK293) protein contents are 24.75±2.32ng and 67.21±2.10ng, and the Benzonase residue contents are 1.55±0.10pg and 1.95±0.16ng per 10(13) vector genome copies (G.C.) separated by CHT/ANX and CsCl. In addition, CHT/ANX yields 798.44±50.10pg of plasmid DNA and 2.17±0.11ng of HEK293 DNA, while CsCl purification yields 840.27±76.14pg of plasmid DNA and 2.43±0.19 of HEK293 DNA. The two methods produce vectors with similar in vitro and in vivo potencies. The results indicated that the CHT/ANX method is suitable for the scalable purification of the rAAV9 vector.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatit, powder, synthetic
Sigma-Aldrich
Calciumphosphat tribasisch, 34.0-40.0% Ca basis
Sigma-Aldrich
Hydroxyapatit, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxylapatit, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxylapatit, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxylapatit, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxylapatit, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Calciumphosphat tribasisch, BioReagent, suitable for plant cell culture, powder