Merck
Role of chlorhexidine in bond strength to artificially eroded dentin over time.
The journal of adhesive dentistry (2015-04-23)
Luciana Fávaro Francisconi-dos-Rios, Leslie Caroll Casas-Apayco, Marcela Pagani Calabria, Paulo Afonso Silveria Francisconi, Ana Flávia Sanches Borges, Linda Wang
ZUSAMMENFASSUNG
To assess the long-term effect of a 2% aqueous chlorhexidine (CHX) solution on bond strength to artificially eroded dentin compared to sound dentin. Flat mid-coronal dentin surfaces of extracted third molars (n = 28) were subjected only to grinding with a 600-grit SiC paper for 1 min (sound dentin S, n = 14) or additionally to erosive pH cycling with a cola-based soft-drink (eroded dentin E, n = 14). After acid etching, rinsing, and air drying, S and E were rehydrated with 1.5 μl of 2% CHX (S2%, n = 7; E2%, n = 7) or of distilled water (control SC, n = 7; EC, n = 7). Composite buildups were incrementally constructed with Filtek Z350 following Adper Single Bond 2 application. Specimens were sectioned into beams, which were subjected to microtensile testing immediately or after 6 or 12 months of aging. Fractured surfaces were observed under a digital microscope (50X magnification). Microtensile bond strength (μTBS) (MPa) was analyzed by three-way ANOVA and Tukey's tests (α = 0.05) and failure mode by the Kruskal-Wallis test (α = 0.05). Compared to sound dentin, eroded dentin was consistently related to lower μTBS. Immediately and after 12-month aging, the effect of CHX was insignificant, but it was significant after 6-month aging, when it conserved the bond strength to both eroded and sound dentin. The percentage of adhesive and mixed failures were equivalent, and significantly more frequent than cohesive failures, whether in dentin or in composite. The 2% CHX effect on bond strength conservation to both eroded and sound dentin was not found to be persistent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Actidion, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximid -Lösung, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Cycloheximid, ≥95% (HPLC)
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Actidion, Biotechnology Performance Certified
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphorsäure-16O4 -Lösung, 70 wt. % in D2O, 99.9 atom % 16O