Merck
  • A novel quantification strategy of transferrin and albumin in human serum by species-unspecific isotope dilution laser ablation inductively coupled plasma mass spectrometry (ICP-MS).
A novel quantification strategy of transferrin and albumin in human serum by species-unspecific isotope dilution laser ablation inductively coupled plasma mass spectrometry (ICP-MS).
Analytica chimica acta (2015-06-16)
Liuxing Feng, Dan Zhang, Jun Wang, Dairui Shen, Hongmei Li
ZUSAMMENFASSUNG
Species-specific (SS) isotope dilution analysis with gel electrophoresis (GE)-laser ablation (LA)-ICP-MS is a promising technique for the quantification of particular metal-binding proteins in biological samples. However, unavailable isotopically enriched spike and metal losses in GE separation are main limitations for SS-isotope dilution PAGE-LA-ICP-MS. In this study, we report for the first time the absolute quantification of transferrin (Tf) and albumin (Alb) in human serum by non-denaturing (native) GE combined with species-unspecific isotope dilution mass spectrometry (IDMS). In order to achieve a homogeneous distribution of both protein and isotope-enriched spike (simulated isotope equilibration), immersing the protein strips with (34)S spike solution after gel electrophoresis was demonstrated to be an effective way of spike addition. Furthermore, effects of immersion time and (34)S spike concentration were investigated to obtain optimal conditions of the post-electrophoresis isotope dilution method. The relative mass of spike and ablated sample (m(sp)/m(sam)) in IDMS equation was calculated by standard Tf and Alb proteins, which could be applied to the quantification of Tf and Alb in ERM-DA470k/IFCC for method confirmation. The results were in agreement with the certified value with good precision and small uncertainty (1.5-3%). In this method, species-specific spike protein is not necessary and the integrity of the heteroatom-protein could be maintained in sample preparation process. Moreover, the application of species-unspecific isotope dilution GE-LA-ICP-MS has the potential to offer reliable, direct and simultaneous quantification of proteins after conventional 1D and 2D gel electrophoretic separations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammoniumpersulfat, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Trizma® Base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%