Merck
  • The role of electron donors generated from UV photolysis for accelerating pyridine biodegradation.
The role of electron donors generated from UV photolysis for accelerating pyridine biodegradation.
Biotechnology and bioengineering (2015-04-10)
Yingxia Tang, Yongming Zhang, Ning Yan, Rui Liu, Bruce E Rittmann
ZUSAMMENFASSUNG
Employing an internal circulation baffled biofilm reactor (ICBBR), we evaluated the mechanisms by which photolysis accelerated the biodegradation and mineralization of pyridine (C5 H5 N), a nitrogen-containing heterocyclic compound. We tested the hypothesis that pyridine oxidation is accelerated because a key photolysis intermediate, succinate, is as electron donor that promotes the initial mono-oxygenation of pyridine. Experimentally, longer photolysis time generated more electron-donor products (succinate), which stimulated faster pyridine biodegradation. This pattern was confirmed by directly adding succinate, and the stimulation effect occurred similarly with addition of the same equivalents of acetate and formate. Succinate, whether generated by UV photolysis or added directly, also accelerated mono-oxygenation of the first biodegradation intermediate, 2-hydroxyl pyridine (2HP). 2HP and pyridine were mutually inhibitory in that their mono-oxygenations competed for internal electron donor; thus, the addition of any readily biodegradable donor accelerated both mono-oxygenation steps, as well as mineralization.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Pyridin, ≥99%