Merck
  • Regulatory Role of Gonadotropins and Local Factors Produced by Ovarian Follicles on In Vitro Resistin Expression and Action on Porcine Follicular Steroidogenesis.
Regulatory Role of Gonadotropins and Local Factors Produced by Ovarian Follicles on In Vitro Resistin Expression and Action on Porcine Follicular Steroidogenesis.
Biology of reproduction (2015-05-01)
Agnieszka Rak, Eliza Drwal, Anna Karpeta, Ewa Ł Gregoraszczuk
ZUSAMMENFASSUNG
Resistin, a hormone secreted by adipocytes, is thought to be important in reproduction. Our previous study demonstrated resistin expression in porcine ovarian follicles and its direct effect on steroidogenesis. The aim of the current study was to evaluate the effect of gonadotropins and the local ovarian factors, such as insulin-like growth factor type 1 (IGF1) and steroids (progesterone, testosterone, and 17 beta-estradiol), on the expression and secretion of resistin, as well as its steroidogenic action. Porcine ovarian follicles were exposed to follicle-stimulating hormone (FSH) and luteinizing hormone (LH) at 50-150 ng/ml, IGF1 (10-100 ng/ml), and steroids at 10(-8) to 10(-6) M for 24 h. Then, mRNA, protein expression, and medium concentration of resistin were determined using real-time PCR, Western blot analysis, and ELISA, respectively. In the subsequent experiments, ovarian follicles were exposed to resistin and/or FSH, LH, IGF1, and steroids, and ovarian steroidogenesis was analyzed. Additionally, we examined the direct effect of resistin on the protein expression of receptors for gonadotropins and investigated local factors. The results showed that gonadotropins and steroids have stimulatory effects but that IGF1 has an inhibitory effect on resistin expression and secretion. Resistin decreased gonadotropins and local hormone-induced steroid secretion and inhibited 3beta-hydroxysteroid dehydrogenase, 17beta-hydroxysteroid dehydrogenase, and cytochrome P450 aromatase protein expression. Additionally, we demonstrated that resistin increased the expression of receptors for progesterone and testosterone. These findings all show that the expression and function of resistin are regulated by gonadotropins and local factors produced by ovarian follicles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamin, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Insulin-like Growth Factor-I human, IGF-I, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
1-Brom-3-chlorpropan, for isolation of RNA
Sigma-Aldrich
Bromphenolblau, titration: suitable
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Ethylendiamin, purified by redistillation, ≥99.5%
Sigma-Aldrich
1-Brom-3-chlorpropan, 99%
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
IGF-I human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Ethylendiamin, BioXtra
Sigma-Aldrich
Ethylendiamin -Lösung, technical, 75-80%
Sigma-Aldrich
Ethylendiamin, meets USP testing specifications
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri