Merck
Application of Plant Extracts for the Prevention of Dental Erosion: An in situ/in vitro Study.
Caries research (2015-08-01)
Marie-Theres Weber, Matthias Hannig, Sandra Pötschke, Franziska Höhne, Christian Hannig
ZUSAMMENFASSUNG
Antiadherent and antibacterial effects of certain plant extracts have been proven to be beneficial in preventive dentistry. In the present in situ/in vitro crossover study, the impact of plant extracts rich in polyphenols on the erosion-protective properties of the in situ pellicle was evaluated. Individual splints were prepared for 12 subjects for intraoral exposure of bovine enamel specimens. Following formation of a 1-min pellicle, watery plant extracts (leaves of the wild form of Ribes nigrum, the wild form of Origanum as well as a combination of both) were administered for 10 min in situ. Alternatively, a mouth rinse with fluorides (Elmex Kariesschutz) was performed for 1 min. After further oral exposure for 19/28 min, respectively, slabs were removed and incubated with HCl in vitro over 120 s (pH 2, 2.3, 3). The resulting calcium and phosphate release was quantified photometrically. Slabs with and without a 30-min in situ pellicle served as controls. The modification of pellicle ultrastructure was evaluated by transmission electron microscopy (TEM). Plant extracts modulated the erosion-protective properties of the native in situ pellicle in all test groups in a pH-dependent manner. The combination of R. nigrum leaves and Origanum enhanced the protective properties of the pellicle at all pH values; the administration of this preparation was comparable, yet superior, to the effect of the fluoridated mouth rinse. TEM images indicated that rinsing with R. nigrum leaves/Origanum yielded a distinctly thicker and more electron-dense pellicle. The combination of certain plant extracts offers a novel approach to the complementary prevention of dental erosion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Osmiumtetroxid, ReagentPlus®, 99.8%
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Osmiumtetroxid -Lösung, 4 wt. % in H2O
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Osmiumtetroxid -Lösung, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Osmiumtetroxid, Sealed ampule.
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Osmiumtetroxid, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphorsäure-16O4 -Lösung, 70 wt. % in D2O, 99.9 atom % 16O