Merck
  • Inflammasome Inhibition Suppresses Alveolar Cell Permeability Through Retention of Neuregulin-1 (NRG-1).
Inflammasome Inhibition Suppresses Alveolar Cell Permeability Through Retention of Neuregulin-1 (NRG-1).
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-07-24)
Rajanbabu Venugopal, Lakshmi Galam, Ruan Cox, Jutaro Fukumoto, Young Cho, Prasanna Tamarapu Parthasarathy, Richard F Lockey, Narasaiah Kolliputi
ZUSAMMENFASSUNG
Neuregulin (NRG)-1-human epidermal receptor (HER)-2 signaling pathway is a key regulator of IL-1β-mediated pulmonary inflammation and epithelial permeability. The inflammasome is a newly discovered molecular platform required for caspase-1 activation and maturation of IL-1β. However, the role of the inflammasome in NRG-1-HER2 signaling-mediated alveolar cell permeability is unknown. The inflammasome was activated or inhibited in THP-1 cells; supernatants from these cells were added to A549 cells and human small airway epithelial cells (HSAEC). The protein expression of NRG-1 and phospho-HER2 (pHER2) were measured by Western blot analysis and epithelial permeability was measured using Lucifer yellow dye. Results reveal that alveolar permeability in A549 cells and HSAEC is increased when treated with supernatants of inflammasome-activated THP-1 cells. Alveolar permeability is significantly suppressed when treated with supernatant of inflammasome-inhibited THP-1 cells. Inflammasome-mediated permeability is decreased when A549 cells and HSAEC are pretreated with IL-1β receptor antagonist (IL-1βRA). In addition, HER2 kinase inhibitor AG825 or NRG-1 inhibitor TAPI inhibits inflammasome-mediated permeability in A549 cells and HSAEC demonstrating critical roles of IL-1β, NRG-1, and HER2 in inflammasome-mediated alveolar permeability. These findings suggest that inflammasome-induced alveolar cell permeability is mediated by NRG-1/HER2 signaling through IL-1β regulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Kaliumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Kaliumchlorid, BioXtra, ≥99.0%
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Kaliumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Kaliumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Glybenclamid, ≥99% (HPLC)
Sigma-Aldrich
IL-1 beta human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Kaliumchlorid -Lösung, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Kaliumchlorid, ≥99.99% trace metals basis
Sigma-Aldrich
Glyburid, meets USP testing specifications
Sigma-Aldrich
Kaliumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Kaliumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis