Merck
  • Synthesis of hollow cobalt oxide nanopowders by a salt-assisted spray pyrolysis process applying nanoscale Kirkendall diffusion and their electrochemical properties.
Synthesis of hollow cobalt oxide nanopowders by a salt-assisted spray pyrolysis process applying nanoscale Kirkendall diffusion and their electrochemical properties.
Physical chemistry chemical physics : PCCP (2015-11-17)
Hyeon Seok Ju, Jung Sang Cho, Jong Hwa Kim, Yun Ju Choi, Yun Chan Kang
ZUSAMMENFASSUNG
A new concept for preparing hollow metal oxide nanopowders by salt-assisted spray pyrolysis applying nanoscale Kirkendall diffusion is introduced. The composite powders of metal oxide and indecomposable metal salt are prepared by spray pyrolysis. Post-treatment under a reducing atmosphere and subsequent washing using distilled water produce aggregation-free metal nanopowders. The metal nanopowders are then transformed into metal oxide hollow nanopowders by nanoscale Kirkendall diffusion. Co3O4 hollow nanopowders are prepared as first target materials. A cobalt oxide-NaCl composite powder prepared by spray pyrolysis transforms into several Co3O4 hollow nanopowders by several treatment processes. The discharge capacities of the Co3O4 nanopowders with filled and hollow structures at a current density of 1 A g(-1) for the 150th cycle are 605 and 775 mA h g(-1), respectively. The hollow structure formed by nanoscale Kirkendall diffusion improves the lithium-ion storage properties of Co3O4 nanopowders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cobalt(II)-nitrat Hexahydrat, ACS reagent, ≥98%
Sigma-Aldrich
Cobalt(II)-nitrat Hexahydrat, reagent grade, 98%
Sigma-Aldrich
Cobalt(II)-nitrat Hexahydrat, 99.999% trace metals basis