Merck
  • Synthesis of water-soluble silicon-porphyrin: protolytic behaviour of axially coordinated hydroxy groups.
Synthesis of water-soluble silicon-porphyrin: protolytic behaviour of axially coordinated hydroxy groups.
Dalton transactions (Cambridge, England : 2003) (2015-11-04)
Sebastian Nybin Remello, Fazalurahman Kuttassery, Takehiro Hirano, Yu Nabetani, Daisuke Yamamoto, Satomi Onuki, Hiroshi Tachibana, Haruo Inoue
ZUSAMMENFASSUNG
A new water-soluble silicon(IV)-tetra(4-carboxyphenyl)porphyrin (SiTCPP) with silicon(iv), the second most abundant element on Earth, in the center of porphyrin was synthesized. Fundamental properties including protolytic behaviour of axially coordinating hydroxy groups, and electrochemical behaviour were characterized. The properties were compared with those of silicon(IV)-tetra(2,4,6-trimethylphenyl)porphyrin (SiTMP) and silicon(IV)-tetra(4-trifluoromethylphenyl)porphyrin (SiTFMPP) and discussed in respect to the electron donating/withdrawing effect of the substituents. Two axially coordinating hydroxy groups of SiTCPP exhibit a four-step protolytic behaviour under the acidic conditions along with a single step protolysis of peripheral carboxyl groups. Though SiTCPP and SiTFMPP did not show any reactivity in the photochemical oxygenation of a substrate with K2PtCl6 as a sacrificial electron acceptor, the first oxidation wave in the electrochemical process of SiTCPP and SiTFMPP showed catalytic behaviour in aqueous acetonitrile solution at any pH condition, in contrast to SiTMP which has only a reversible oxidation wave under neutral and weakly acidic conditions. The criteria for the electrochemical oxidative activation of water and the photooxygenation of the substrate were obtained. The higher oxidation wave of Si-porphyrins than ∼0.86 volt vs. SHE is required for the electrochemical oxidation of water, while suitable protecting groups such as a methyl substituent is a requisite for the photochemical oxygenation with K2PtCl6 as a sacrificial electron acceptor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithium-bis-(trimethylsilyl)-amid -Lösung, 1.0 M in THF
Sigma-Aldrich
1,2-Dimethoxyethan, ReagentPlus®, ≥99%, inhibitor-free
Sigma-Aldrich
1,2-Dimethoxyethan, anhydrous, 99.5%, inhibitor-free
Sigma-Aldrich
Lithium-bis-(trimethylsilyl)-amid, 97%
Sigma-Aldrich
Trichlorosilan, 99%
Sigma-Aldrich
Lithium-bis-(trimethylsilyl)-amid -Lösung, 1 M in toluene
Sigma-Aldrich
Lithium-bis-(trimethylsilyl)-amid -Lösung, 1.5 M in THF
Sigma-Aldrich
Cyclohexen, inhibitor-free, ReagentPlus®, 99%
Sigma-Aldrich
Lithium-bis-(trimethylsilyl)-amid -Lösung, 1.0 M in hexanes
Sigma-Aldrich
Cyclohexen, contains 100 ppm BHT as inhibitor, ≥99.0%
Sigma-Aldrich
Lithium-bis-(trimethylsilyl)-amid -Lösung, 0.5 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium-bis-(trimethylsilyl)-amid -Lösung, 1 M in tert-butyl methyl ether