Merck
Inferring interaction type in gene regulatory networks using co-expression data.
Algorithms for molecular biology : AMB (2015-07-15)
Pegah Khosravi, Vahid H Gazestani, Leila Pirhaji, Brian Law, Mehdi Sadeghi, Bahram Goliaei, Gary D Bader
ZUSAMMENFASSUNG
Knowledge of interaction types in biological networks is important for understanding the functional organization of the cell. Currently information-based approaches are widely used for inferring gene regulatory interactions from genomics data, such as gene expression profiles; however, these approaches do not provide evidence about the regulation type (positive or negative sign) of the interaction. This paper describes a novel algorithm, "Signing of Regulatory Networks" (SIREN), which can infer the regulatory type of interactions in a known gene regulatory network (GRN) given corresponding genome-wide gene expression data. To assess our new approach, we applied it to three different benchmark gene regulatory networks, including Escherichia coli, prostate cancer, and an in silico constructed network. Our new method has approximately 68, 70, and 100 percent accuracy, respectively, for these networks. To showcase the utility of SIREN algorithm, we used it to predict previously unknown regulation types for 454 interactions related to the prostate cancer GRN. SIREN is an efficient algorithm with low computational complexity; hence, it is applicable to large biological networks. It can serve as a complementary approach for a wide range of network reconstruction methods that do not provide information about the interaction type.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridiniumchlorchromat, 98%