Merck
  • Determination of terpenoid indole alkaloids in hairy roots of Rhazya stricta (Apocynaceae) by GC-MS.
Determination of terpenoid indole alkaloids in hairy roots of Rhazya stricta (Apocynaceae) by GC-MS.
Phytochemical analysis : PCA (2015-06-23)
Amir Akhgari, Into Laakso, Tuulikki Seppänen-Laakso, Teijo Yrjönen, Heikki Vuorela, Kirsi-Marja Oksman-Caldentey, Heiko Rischer
ZUSAMMENFASSUNG
Rhazya stricta Decne. (Apocynaceae) is a medicinal plant rich in terpenoid indole alkaloids (TIAs), some of which possess important pharmacological properties. The study material including transgenic hairy root cultures have been developed and their potential for alkaloid production are being investigated. In this study, a comprehensive GC-MS method for qualitative and quantitative analysis of alkaloids from Rhazya hairy roots was developed. The composition of alkaloids was determined by using GC-MS. In quantification, the ratio between alkaloid and internal standard was based on extracted ion from total ion current (TIC) analyses. The developed method was validated. An acceptable precision with RSD ≤ 8% over a linear range of 1 to 100 µg/mL was achieved. The accuracy of the method was within 94-107%. Analysis of hairy root extracts indicated the occurrence of a total of 20 TIAs. Six of them, pleiocarpamine, fluorocarpamine, vincamine, ajmalicine and two yohimbine isomers are reported here for the first time in Rhazya. Trimethylsilyl (TMS) derivatisation of the extracts resulted in the separation of two isomers for yohimbine and also for vallesiachotamine. Clearly improved chromatographic profiles of TMS-derivatives were observed for vincanine and for minor compounds vincamine and rhazine. The results show that the present GC-MS method is reliable and well applicable for studying the variation of indole alkaloids in Rhazya samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Essigsäureanhydrid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
N-Methyl-N-trimethylsilyltrifluoracetamid, synthesis grade
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Essigsäureanhydrid, ACS reagent, ≥98.0%
Supelco
N-Methyl-N-trimethylsilyltrifluoracetamid, BioReagent, for silylations, LiChropur
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%
Sigma-Aldrich
Essigsäureanhydrid, 99.5%
Sigma-Aldrich
Ammoniak -Lösung, 4 M in methanol
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in THF
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Supelco
N-Methyl-bis-trifluoracetamid, derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Yohimbin -hydrochlorid, ≥98% (HPLC), powder
Sigma-Aldrich
Essigsäureanhydrid, Arxada quality, ≥99.5% (GC)
Sigma-Aldrich
Ammoniak-14N, 99.99 atom % 14N