Merck
  • Distribution of radioactive cesium and stable cesium in cattle kept on a highly contaminated area of Fukushima nuclear accident.
Distribution of radioactive cesium and stable cesium in cattle kept on a highly contaminated area of Fukushima nuclear accident.
Animal science journal = Nihon chikusan Gakkaiho (2014-12-17)
Itaru Sato, Keiji Okada, Jun Sasaki, Hiroyuki Chida, Hiroshi Satoh, Kiyoshi Miura, Kaoru Kikuchi, Kumiko Otani, Shusuke Sato
ZUSAMMENFASSUNG
Radioactivity inspection of slaughtered cattle is generally conducted using a portion of the neck muscle; however, there is limited information about the distribution of radioactive cesium in cattle. In this study, therefore, we measured not only radioactive cesium but also stable cesium in various tissues of 19 cattle that had been kept in the area highly contaminated by the Fukushima nuclear accident. Skeletal muscles showed approximately 1.5-3.0 times higher concentration of radioactive cesium than internal organs. Radioactive cesium concentration in the tenderloin and top round was about 1.2 times as high as that in the neck muscle. The kidney showed the highest concentration of radioactive cesium among internal organs, whereas the liver was lowest. Radioactive cesium concentration in the blood was about 8% of that in the neck muscle. Characteristics of stable cesium distribution were almost the same as those of radioactive cesium. Correlation coefficient between radioactive cesium and stable cesium in tissues of individual cattle was 0.981 ± 0.012. When a suspicious level near 100 Bq/kg is detected in the neck of slaughtered cattle, re-inspection should be conducted using a different region of muscle, for example top round, to prevent marketing of beef that violates the Food Sanitation Act.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumiodid, 99.999% trace metals basis
Sigma-Aldrich
Natriumiodid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumiodid, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumiodid, AnhydroBeads, −10 mesh, 99.999% trace metals basis