Merck
Convenient surface functionalization of whole-Teflon chips with polydopamine coating.
Biomicrofluidics (2015-09-05)
Bo Shen, Bin Xiong, Hongkai Wu
ZUSAMMENFASSUNG
This paper presents a convenient strategy to modify the surface of whole-Teflon microfluidic chips by coating the channel walls with a thin layer of polydopamine (PDA) film, which is formed by oxidation-induced self-polymerization of dopamine in alkaline solution. Two coating strategies, static incubation and dynamic flow, are demonstrated and used for tuning the physical and chemical properties of the coated channel walls. The functionalized surfaces were investigated with the contact angle, X-ray photoelectron spectroscopy, and atomic force microscopy measurements. The coating time was optimized according to the fluorescent intensity of the green fluorescent protein immobilized on the modified surface. Applications of the PDA-modified Teflon microchips in bioanalysis were demonstrated with a typical sandwich immunoassay. Moreover, long-term cell culture experiments on modified and native Teflon chips revealed that the chip biocompatibility can be greatly improved with PDA coating. The results indicate that the surface properties of the Teflon can be easily controlled by the PDA modification, thus greatly expanding the application scope of whole-Teflon chips for various chemical and biological research fields.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)