Merck
  • Cortical oscillatory dynamics and benzodiazepine-site modulation of tonic inhibition in fast spiking interneurons.
Cortical oscillatory dynamics and benzodiazepine-site modulation of tonic inhibition in fast spiking interneurons.
Neuropharmacology (2015-03-24)
Emma J Prokic, Cathryn Weston, Naoki Yamawaki, Stephen D Hall, Roland S G Jones, Ian M Stanford, Graham Ladds, Gavin L Woodhall
ZUSAMMENFASSUNG
Tonic conductance mediated by extrasynaptic GABAA receptors has been implicated in the modulation of network oscillatory activity. Using an in vitro brain slice to produce oscillatory activity and a kinetic model of GABAA receptor dynamics, we show that changes in tonic inhibitory input to fast spiking interneurons underlie benzodiazepine-site mediated modulation of neuronal network synchrony in rat primary motor cortex. We found that low concentrations (10 nM) of the benzodiazepine site agonist, zolpidem, reduced the power of pharmacologically-induced beta-frequency (15-30 Hz) oscillatory activity. By contrast, higher doses augmented beta power. Application of the antagonist, flumazenil, also increased beta power suggesting endogenous modulation of the benzodiazepine binding site. Voltage-clamp experiments revealed that pharmacologically-induced rhythmic inhibitory postsynaptic currents were reduced by 10 nM zolpidem, suggesting an action on inhibitory interneurons. Further voltage-clamp studies of fast spiking cells showed that 10 nM zolpidem augmented a tonic inhibitory GABAA receptor mediated current in fast spiking cells whilst higher concentrations of zolpidem reduced the tonic current. A kinetic model of zolpidem-sensitive GABAA receptors suggested that incubation with 10 nM zolpidem resulted in a high proportion of GABAA receptors locked in a kinetically slow desensitized state whilst 30 nM zolpidem favoured rapid transition into and out of desensitized states. This was confirmed experimentally using a challenge with saturating concentrations of GABA. Selective modulation of an interneuron-specific tonic current may underlie the reversal of cognitive and motor deficits afforded by low-dose zolpidem in neuropathological states.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
(+)-Bicucullin, ≥97.0% (TLC)
Sigma-Aldrich
Flumazenil, >99% (HPLC), solid